Search
Search

glod5| Gene Symbol: glod5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045618.1 | NP_001039083.1 | 3 - 4 | 355 | 91 | |
| GenBank mRNA | BC135158.1 | - | 3 - 4 | 369 | 91 | |
| CR926243.1 | - | 3 - 4 | 355 | 91 | ||
Set Membership: |
|