Search
Search

bmp1| Gene Symbol: bmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045671.1 | NP_001039136.1 | 12 - 13 | 1629 | 65 | |
| XM_012959185.2 | XP_012814639.1 | 12 - 13 | 1721 | 65 | ||
| GenBank mRNA | CR942509.2 | - | 12 - 13 | 1665 | 65 | |
Set Membership: |
|