Search
Search

kiaa1109| Gene Symbol: kiaa1109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078735.1 | NP_001072203.1 | 12 - 13 | 1475 | 67 | |
| GenBank mRNA | BC118757.1 | - | 12 - 13 | 1475 | 67 | |
Set Membership: |
|