Search
Search

acsl6| Gene Symbol: acsl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078862.1 | NP_001072330.1 | 15 - 16 | 1726 | 101 | |
| XM_012959214.2 | XP_012814668.1 | 15 - 16 | 1956 | 101 | ||
| XM_012959215.2 | XP_012814669.1 | 15 - 16 | 1958 | 101 | ||
| XM_012959216.1 | XP_012814670.1 | 15 - 16 | 1745 | 101 | ||
| GenBank mRNA | BC121404.1 | - | 15 - 16 | 1726 | 101 | |
Set Membership: |
|