Search
Search

zfat| Gene Symbol: zfat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079573.1 | NP_001073041.1 | 2 - 3 | 290 | 64 | |
| XM_012964853.2 | XP_012820307.1 | 2 - 3 | 372 | 64 | ||
| GenBank mRNA | BC121281.1 | - | 2 - 3 | 290 | 64 | |
Set Membership: |
|