Search
Search

cnot1| Gene Symbol: cnot1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097189.1 | NP_001090658.1 | 33 - 34 | 5053 | 98 | |
| XM_018092858.1 | XP_017948347.1 | 32 - 33 | 4999 | 98 | ||
| GenBank mRNA | BC127296.1 | - | 33 - 34 | 5053 | 98 | |
Set Membership: |
|