Search
Search

fbxo11| Gene Symbol: fbxo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012963878.2 | XP_012819332.1 | 2 - 3 | 2126 | 72 | |
| XM_012963879.2 | XP_012819333.1 | 2 - 3 | 2127 | 72 | ||
| XM_018094133.1 | XP_017949622.1 | 2 - 3 | 241 | 72 | ||
| XM_018094134.1 | XP_017949623.1 | 2 - 3 | 2131 | 72 | ||
| GenBank mRNA | BC127360.1 | - | 2 - 3 | 458 | 72 | |
| CT030543.1 | - | 2 - 3 | 306 | 72 | ||
Set Membership: |
|