Search
Search

secisbp2| Gene Symbol: secisbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097262.1 | NP_001090731.1 | 8 - 9 | 1078 | 89 | |
| GenBank mRNA | BC128999.1 | - | 8 - 9 | 1078 | 89 | |
Set Membership: |
|