Search
Search

pank3| Gene Symbol: pank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113906.1 | NP_001107378.1 | 6 - 7 | 1405 | 77 | |
| GenBank mRNA | BC157250.1 | - | 6 - 7 | 1405 | 77 | |
| BC170962.1 | - | 6 - 7 | 1102 | 77 | ||
| BC170964.1 | - | 6 - 7 | 1102 | 77 | ||
Set Membership: |
|