Search
Search

zcrb1| Gene Symbol: zcrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114501.2 | NP_001107973.1 | 6 - 7 | 523 | 75 | |
| XM_012958532.2 | XP_012813986.1 | 7 - 8 | 615 | 75 | ||
| GenBank mRNA | BC158310.1 | - | 6 - 7 | 525 | 75 | |
| CU025042.1 | - | 7 - 8 | 550 | 75 | ||
Set Membership: |
|