Search
Search

ptpn6| Gene Symbol: ptpn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123456.1 | NP_001116928.1 | 14 - 15 | 1874 | 88 | |
| GenBank mRNA | BC160526.1 | - | 14 - 15 | 1874 | 88 | |
Set Membership: |
|