Search
Search

pbx2| Gene Symbol: pbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128661.1 | NP_001122133.1 | 8 - 9 | 1625 | 77 | |
| XM_012967841.2 | XP_012823295.1 | 7 - 8 | 1596 | 77 | ||
| GenBank mRNA | BC135908.1 | - | 8 - 9 | 1556 | 77 | |
| BC167699.1 | - | 8 - 9 | 1625 | 77 | ||
Set Membership: |
|