Search
Search

zmiz1| Gene Symbol: zmiz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142079.1 | NP_001135551.1 | 16 - 17 | 2598 | 63 | |
| XM_012966212.2 | XP_012821666.1 | 23 - 24 | 3262 | 63 | ||
| XM_012966213.2 | XP_012821667.1 | 22 - 23 | 3208 | 63 | ||
| XM_012966214.2 | XP_012821668.1 | 25 - 26 | 3496 | 63 | ||
| XM_012966215.2 | XP_012821669.1 | 24 - 25 | 3430 | 63 | ||
| XM_012966216.2 | XP_012821670.1 | 24 - 25 | 3422 | 63 | ||
| XM_012966217.2 | XP_012821671.1 | 24 - 25 | 3412 | 63 | ||
| XM_012966219.2 | XP_012821673.1 | 23 - 24 | 3352 | 63 | ||
| XM_012966220.2 | XP_012821674.1 | 16 - 17 | 2661 | 63 | ||
| XM_012966221.2 | XP_012821675.1 | 16 - 17 | 2996 | 63 | ||
| XM_018095300.1 | XP_017950789.1 | 24 - 25 | 3439 | 63 | ||
| XM_018095301.1 | XP_017950790.1 | 23 - 24 | 3385 | 63 | ||
| XM_018095302.1 | XP_017950791.1 | 24 - 25 | 3324 | 63 | ||
| XM_018095303.1 | XP_017950792.1 | 23 - 24 | 3151 | 63 | ||
| XM_018095304.1 | XP_017950793.1 | 23 - 24 | 3361 | 63 | ||
| XM_018095305.1 | XP_017950794.1 | 17 - 18 | 2774 | 63 | ||
| GenBank mRNA | BC168031.1 | - | 16 - 17 | 2598 | 63 | |
Set Membership: |
|