Search
Search

LOC100281278| Gene Symbol: LOC100281278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154197.1 | NP_001147669.1 | 602 | 81 | ||
| GenBank mRNA | BT068804.1 | - | 545 | 81 | ||
| BT087798.1 | - | 592 | 81 | |||
| EU956206.1 | - | 602 | 81 | |||
Set Membership: |
|