Search
Search

LOC100272652| Gene Symbol: LOC100272652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001147110.1 | NP_001140582.1 | 324 | 74 | ||
| GenBank mRNA | BT039113.1 | - | 324 | 74 | ||
| JN634078.1 | - | 228 | 74 | |||
Set Membership: |
|