Search
Search

LOC100273326| Gene Symbol: LOC100273326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001147767.1 | NP_001141239.1 | 402 | 84 | ||
| GenBank mRNA | BT040816.1 | - | 402 | 84 | ||
| EU954504.1 | - | 364 | 84 | |||
| EU966100.1 | - | 364 | 84 | |||