Search
Search

LOC100284204| Gene Symbol: LOC100284204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157099.1 | NP_001150571.1 | 327 | 63 | ||
| GenBank mRNA | EU959446.1 | - | 287 | 63 | ||
| EU967491.1 | - | 327 | 63 | |||
Set Membership: |
|