Search
Search

LOC541833| Gene Symbol: LOC541833 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111514.1 | NP_001104984.1 | 188 | 121 | ||
| GenBank mRNA | AF244683.1 | - | 188 | 121 | ||
| EU966577.1 | - | 234 | 121 | |||
Set Membership: |
|