Search
Search

cl12955_1| Gene Symbol: cl12955_1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136602.1 | NP_001130074.1 | 439 | 72 | ||
| GenBank mRNA | BT033191.1 | - | 389 | 72 | ||
| BT083482.1 | - | 363 | 72 | |||
| EU970898.1 | - | 439 | 72 | |||
Set Membership: |
|