Search
Search

LOC100216983| Gene Symbol: LOC100216983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143362.1 | NP_001136834.1 | 385 | 83 | ||
| GenBank mRNA | BT037727.1 | - | 385 | 83 | ||
| EU970271.1 | - | 289 | 83 | |||
Set Membership: |
|