Search
Search

LOC100284946| Gene Symbol: LOC100284946 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157841.1 | NP_001151313.1 | 1234 | 104 | ||
| GenBank mRNA | EU970234.1 | - | 1234 | 104 | ||
Set Membership: |
|