Search
SNP Genotyping Assays |
Single nucleotide polymorphisms (SNPs) are a common source of genetic variation within genomic DNA and typically confer a single nucleotide difference within a specific gene locus, resulting in two alleles of the gene. SNPs within the coding or regulatory regions of a gene can affect function, or more commonly serve as biological markers for inheritance of specific loci. TaqMan SNP Genotyping Assays help detect and distinguish SNPs accurately and efficiently, so that their associations and roles in gene function and disease can be better understood.
Thermo Fisher Scientific’s collection of predesigned TaqMan assays feature over 17 million SNP assays, HapMap SNPs, nearly 1 million coding SNPs, and 1.4 million SNPs from the 1000 Genomes Project database. Our expanded content provides confidence that we have a predesigned assay to help accelerate your human and mouse SNP genotyping studies. Search and order predesigned SNP genotyping assays with the tool below.
Learn more: How to search TaqMan SNP genotyping assays using search tool
Select SNP genotyping assays based on your instrument setup
Each TaqMan SNP Genotyping assay includes two differentially labeled, allele-specific TaqMan MGB probes and a PCR primer pair that uniquely amplify and offer outstanding analytical specificity for the allele of interest. Choose an assay from one of more than 17 million predesigned assays or customize your own using our Custom Assay Design Tool.
Utilize our proprietary assay design algorithm to design a custom SNP genotyping assay while enjoying the benefits of TaqMan chemistry and an easy-to-use workflow.
Order now:Custom TaqMan SNP Genotyping Assay, non-human
Order now:Custom TaqMan SNP Genotyping Assay, human
Order TaqMan SNP genotyping assays pre-plated on 96- or 384-well plates using the TaqMan custom plating service. Customize the experiment to analyze either a few SNPs in many samples or many SNPs in a few samples.
Explore: Flexible SNP genotyping arrays
Supports high sample throughput for mid-density genotyping: >110,000 genotypes in a single day with the QuantStudio 12K Flex Real-Time PCR System.
Like all TaqMan Assays, TaqMan SNP genotyping assays require a double-stranded DNA template, the Taq polymerase enzyme, and two primers—forward and reverse—specific to the sequence to be amplified. Unlike gene expression qPCR, SNP detection requires two probes with different fluorescent reporters to differentiate homozygous from heterozygous samples.
The first probe, labeled with VIC fluorescent dye, detects the first allele sequence, while the second probe, labeled with FAM fluorescent dye, detects the second allele sequence. If the sample is homozygous for allele 1, the fluorescence readout or 'allelic discrimination plot' will show mostly VIC fluorescence. If the sample is homozygous for allele 2, the plot will mainly show signal from the FAM dye. If the sample is heterozygous, there should be roughly equal signal for each dye. Across an entire plate, the data should resolve into three discrete clusters. Exceptions can occur if the minor allele frequency is very low, if there is a copy number variation (CNV) of the gene in question, or if it lies on the X chromosome.
For their TaqMan SNP genotyping assays, the context sequence of the assay is always listed in the forward orientation, regardless of the strand on which the SNP is typically reported. It is important to compare the context sequence of the SNP assay to the SNP sequence in the NCBI’s dbSNP database to determine which dye is associated with the minor allele.
Three key factors contribute to the discrimination of a single base mismatch in genotyping assays:
We provide multiple solutions to isolate and purify DNA.
Kits for genomic DNA extraction
Explore our broad range of kits for purifying genomic DNA from a variety of samples including tissue, cells, blood, serum, plants, forensic samples, and more.
DNA Extract All Reagents Kit
Obtain PCR-ready DNA from a wide variety of sample types, ranging from blood to buccal swabs to plant tissues in five minutes.
We offer a variety of TaqMan Genotyping Assays for several real-time PCR analysis applications.
TaqMan SNP Genotyping Assays
The Predesigned TaqMan SNP Genotyping Assays collection includes millions of genome‑wide human assays, including common 1,000 Genome SNPs, HapMap SNPs, and coding SNP assays.
Custom TaqMan SNP Genotyping Assays
Create custom assays by submitting confidential target sequences to our secure assay design pipeline.
TaqMan Drug Metabolism Assays
A comprehensive collection of inventoried assays optimized for genotyping SNPs, insertions and deletions (indels), and multi‑nucleotide polymorphisms (MNPs) in drug metabolism-related genes.
Prepare a reaction plate using TaqMan Assays and master mix, along with your DNA samples. No additional reagents are needed.
TaqPath ProAmp Master Mix (includes ROX)
TaqPath ProAmp Master Mix (includes MUSTANG PURPLE)- for higher multiplexing, up to 4 targets
Delivers accurate genotyping results, even from samples containing PCR inhibitors.
Amplify DNA and achieve allelic discrimination by running the PCR reaction on an Applied Biosystems real-time PCR system and/or thermal cycler. To better evaluate the accuracy of genotype calls, perform real-time time PCR and obtain genotyping data for every cycle. Or for higher throughput, run PCR on a thermal cycler and then perform an endpoint read on a real-time instrument.
ProFlex PCR System
The ProFlex PCR System’s interchangeable block formats allow you to maximize your throughput or run independent experiments concurrently.
SimpliAmp Thermal Cycler
The SimpliAmp Thermal Cycler is an easy-to use, compact, and accurate thermal cycler designed to fit every lab’s essential PCR workflow.
QuantStudio 3 Real-Time PCR System
Simple to use for all levels of experience
QuantStudio 5 Real-Time PCR System
Easy controls for optimized experiments
QuantStudio 6 Flex Real-Time PCR System
High flexibility and intuitive design
QuantStudio 7 Flex Real-Time PCR System
High versatility and intuitive design
QuantStudio 6 Pro Real-Time PCR System
High throughput, flexibility and smart features
Quantstudio 7 Pro Real-Time PCR System
High throughput, flexibility, automation capability
QuantStudio 12K Flex Real-Time PCR System
Maximum throughput, flexibility, and scalability
Process and interpret qPCR data with intuitive software data analysis tools for a range of applications.
Diomni Design and Analysis (RUO) Software
Available in desktop, cloud, and on-premises configurations, the Diomni Design and Analysis (RUO) Software is a comprehensive solution for plate setup, quality checks, and results review. It supports qPCR analysis with an intuitive user interface, multi-plate analysis, and a proprietary genotyping algorithm. Enhanced visuals and integrated traces for allelic discrimination ensure precise SNP result quality control.
Applied Biosystems qPCR Analysis Modules
Our cloud-based applications can bring together multiple data sets, with analysis speeds up to 10 times faster than our desktop software. The Genotyping app includes improved visuals and integrated traces of allelic discrimination plots to allow thorough QC of SNP assays to accurately reflect the true signals versus background noise.
TaqMan Genotyper Software
TaqMan Genotyper Software is for use with TaqMan SNP Genotyping Assays (Predesigned, Custom & DME) in combination with 48-, 96-, and 384-well microplates or OpenArray Genotyping Plates, and offers a state-of-the-art genotype-calling algorithm, an intuitive user interface, and enhanced multi-plate analysis features.
We provide multiple solutions to isolate and purify DNA.
Kits for genomic DNA extraction
Explore our broad range of kits for purifying genomic DNA from a variety of samples including tissue, cells, blood, serum, plants, forensic samples, and more.
DNA Extract All Reagents Kit
Obtain PCR-ready DNA from a wide variety of sample types, ranging from blood to buccal swabs to plant tissues in five minutes.
We offer a variety of TaqMan Genotyping Assays for several real-time PCR analysis applications.
TaqMan SNP Genotyping Assays
The Predesigned TaqMan SNP Genotyping Assays collection includes millions of genome‑wide human assays, including common 1,000 Genome SNPs, HapMap SNPs, and coding SNP assays.
Custom TaqMan SNP Genotyping Assays
Create custom assays by submitting confidential target sequences to our secure assay design pipeline.
TaqMan Drug Metabolism Assays
A comprehensive collection of inventoried assays optimized for genotyping SNPs, insertions and deletions (indels), and multi‑nucleotide polymorphisms (MNPs) in drug metabolism-related genes.
Prepare a reaction plate using TaqMan Assays and master mix, along with your DNA samples. No additional reagents are needed.
TaqPath ProAmp Master Mix (includes ROX)
TaqPath ProAmp Master Mix (includes MUSTANG PURPLE)- for higher multiplexing, up to 4 targets
Delivers accurate genotyping results, even from samples containing PCR inhibitors.
Amplify DNA and achieve allelic discrimination by running the PCR reaction on an Applied Biosystems real-time PCR system and/or thermal cycler. To better evaluate the accuracy of genotype calls, perform real-time time PCR and obtain genotyping data for every cycle. Or for higher throughput, run PCR on a thermal cycler and then perform an endpoint read on a real-time instrument.
ProFlex PCR System
The ProFlex PCR System’s interchangeable block formats allow you to maximize your throughput or run independent experiments concurrently.
SimpliAmp Thermal Cycler
The SimpliAmp Thermal Cycler is an easy-to use, compact, and accurate thermal cycler designed to fit every lab’s essential PCR workflow.
QuantStudio 3 Real-Time PCR System
Simple to use for all levels of experience
QuantStudio 5 Real-Time PCR System
Easy controls for optimized experiments
QuantStudio 6 Flex Real-Time PCR System
High flexibility and intuitive design
QuantStudio 7 Flex Real-Time PCR System
High versatility and intuitive design
QuantStudio 6 Pro Real-Time PCR System
High throughput, flexibility and smart features
Quantstudio 7 Pro Real-Time PCR System
High throughput, flexibility, automation capability
QuantStudio 12K Flex Real-Time PCR System
Maximum throughput, flexibility, and scalability
Process and interpret qPCR data with intuitive software data analysis tools for a range of applications.
Diomni Design and Analysis (RUO) Software
Available in desktop, cloud, and on-premises configurations, the Diomni Design and Analysis (RUO) Software is a comprehensive solution for plate setup, quality checks, and results review. It supports qPCR analysis with an intuitive user interface, multi-plate analysis, and a proprietary genotyping algorithm. Enhanced visuals and integrated traces for allelic discrimination ensure precise SNP result quality control.
Applied Biosystems qPCR Analysis Modules
Our cloud-based applications can bring together multiple data sets, with analysis speeds up to 10 times faster than our desktop software. The Genotyping app includes improved visuals and integrated traces of allelic discrimination plots to allow thorough QC of SNP assays to accurately reflect the true signals versus background noise.
TaqMan Genotyper Software
TaqMan Genotyper Software is for use with TaqMan SNP Genotyping Assays (Predesigned, Custom & DME) in combination with 48-, 96-, and 384-well microplates or OpenArray Genotyping Plates, and offers a state-of-the-art genotype-calling algorithm, an intuitive user interface, and enhanced multi-plate analysis features.
For Research Use Only. Not for use in diagnostic procedures.