PrepSEQ™ 1-2-3 Nucleic Acid Extraction Kit
Applied Biosystems™

PrepSEQ™ 1-2-3 Nucleic Acid Extraction Kit

The PrepSEQ 1-2-3 Nucleic Acid Extraction Kit uses magnetic particle-based separation technology to extract DNA from Mycoplasma cells or Mouse Minute Virus (MMV) isolated from a variety of starting material.
Have Questions?
Catalog NumberQuantity
4452222100 Preps
Catalog number 4452222
Price (USD)
1,144.00
Each
Quantity:
100 Preps
Price (USD)
1,144.00
Each

The PrepSEQ 1-2-3 Nucleic Acid Extraction Kit uses magnetic particle-based separation technology to extract DNA from Mycoplasma cells or Mouse Minute Virus (MMV) isolated from a variety of starting material, such as infected cell cultures or Mycoplasma liquid cultures.

Using this kit, lysate samples are prepared prior to extracting the target DNA. For samples that contain Mycoplasma, the lysates contain Mycoplasma DNA from the free organisms in the culture media and from organisms released from the infected host cells.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Final Product TypeGenomic DNA, Viral DNA, Bacterial DNA
For Use With (Application)Cell Culture Contamination Detection
Product LinePrepSEQ
Quantity100 Preps
Sample TypeCell Culture Supernatants
TargetGenomic DNA
Target Organism ClassMouse Minute Virus (MMV), Mycoplasma spp., Vesivirus
FormatKit
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage
• PrepSEQ Proteinase K (store at -15°C to -25°C)
• RNASE Cocktail (-15°C to -25°C)
• PrepSEQ Extraction Kit including lysis buffer, Proteinase K buffer, binding solution, wash and elution buffers (room temperature)
• PrepSEQ Magnetic Beads (room temperature)