Instruments and software for polymer and composite characterization
Integrated instruments and software for polymer characterization
Understanding polymer and composite performance requires more than a single analytical technique. Researchers often need to combine structural, compositional, surface, nanoscale, and three-dimensional information to understand how chemistry, processing, morphology, and reinforcement architecture influence material behavior. Our integrated portfolio of microscopy, surface analysis, and visualization technologies supports these workflows and helps researchers investigate materials from the microscale to the nanoscale.
Scanning electron microscopes (SEMs)
Polymer and composite researchers frequently need to understand how morphology, defects, fillers, reinforcements, fiber orientation, and processing conditions influence material performance. Thermo Scientific SEMs provide high-resolution imaging and compositional analysis to help reveal fracture features, phase morphology, filler dispersion, fiber distribution, voids, and manufacturing defects. By combining structural and elemental information within a single workflow, SEMs help researchers investigate the relationships between microstructure and performance across a wide range of polymer and composite materials, including plastics, elastomers, coatings, adhesives, and reinforced composites.
Focused ion beam scanning electron microscopes (FIB-SEMs)
Many critical features in polymer and composite systems exist beneath the surface. Multilayer packaging, adhesive interfaces, coatings, fiber-reinforced composites, laminates, and buried defects often require cross-sectional investigation to fully understand their structure and behavior. Thermo Scientific FIB-SEMs offer precise site-specific sectioning and imaging of internal features, providing valuable insight into interfacial interactions, failure mechanisms, material architecture, void formation, and defect formation. These workflows are particularly valuable for advanced polymer and composite systems where understanding three-dimensional structure is essential.
Transmission electron microscopes (TEMs)
As polymer and composite formulations become increasingly sophisticated, many performance-driving features occur at nanometer length scales. Thermo Scientific TEMs help researchers investigate phase separation, crystallinity, nanoparticle dispersion, interfacial layers, sizing chemistries, and nanostructured materials with exceptional resolution. These capabilities help connect nanoscale architecture with mechanical, thermal, electrical, and functional performance, supporting both fundamental research and advanced materials development.
X-ray photoelectron spectroscopy (XPS) systems
Many polymer and composite properties are governed by surface and interface chemistry rather than bulk composition alone. Adhesion, coating performance, wettability, printability, fiber-matrix bonding, contamination, and environmental durability are often determined by chemical interactions occurring within only a few nanometers of the surface. Thermo Scientific XPS systems provide highly surface-sensitive chemical analysis that helps researchers investigate elemental composition, chemical bonding, oxidation states, and surface modifications. These insights help optimize coatings, adhesives, films, sizing treatments, and surface treatments while improving understanding of interface-driven performance.
Avizo Software
Modern polymer and composite characterization increasingly relies on extracting quantitative information from complex imaging datasets. Thermo Scientific Avizo Software offers advanced visualization, AI-assisted segmentation, and quantitative analysis of two-dimensional and three-dimensional data, helping researchers transform images into actionable insights. By quantifying fiber orientation, phase distribution, porosity, and defects, Avizo Software helps researchers and engineers better understand structure-property relationships and communicate results with greater confidence.
For Research Use Only. Not for use in diagnostic procedures.