Reverse Transcription
We offer a selection of high-quality products for first-strand cDNA synthesis to meet your research needs. Choose from a variety of reverse transcription enzymes, formats, primers, and dNTP solutions that work best for your application. With our premium line of Invitrogen SuperScript Reverse Transcriptases, we have what you need for superior cDNA synthesis results with any type of template RNA.
Reverse transcription products
Stand-alone enzymes for maximum flexibility in reaction setup
  Superior enzyme for all RNAs, including difficult templates Value enzyme for commonly used RNA templates
  Superscript IV Reverse Transcriptase M-MLV-Reverse Transcriptase
Optimal reaction temp 50–55°C 37°C
Reaction time 10 min 50 min
cDNA yield Superior Average
Sensitivity Superior Average
Max cDNA length Up to 12 kb Up to 7 kb
Ability to work with degraded or inhibitor-containing RNA High Low
Reduced RNase H activity Yes No
  Learn more Learn more
Other reverse transcriptases
Complete kits with all cDNA synthesis reaction components
  Superior choice for all RNAs, including difficult templates Value choice for commonly used RNA templates
  SuperScript IV
First-Strand Synthesis System
High-Capacity cDNA Reverse Transcription Kit
Reverse transcriptase SuperScript IV MultiScribe MuLV
Optimal reaction temp 50–55°C 37°C
Reaction time 10 min 50 min
cDNA yield Superior Average
Sensitivity Superior Average
Max cDNA length Up to 12 kb Up to 7 kb
Ability to work with degraded or inhibitor-containing RNA High Low
Reduced RNase H activity Yes No
  Learn more Learn more
Other cDNA synthesis kits for endpoint PCR applications
cDNA synthesis kits for the highest convenience in RT-qPCR setup
  Superior master mix for all RNA templates Superior master mix for all RNA templates, with fast and easy gDNA removal Value choice for commonly used RNA templates
  SuperScript IV VILO Master Mix SuperScript IV VILO Master Mix with ezDNase Enzyme High-Capacity cDNA Reverse Transcription Kit
Reverse transcriptase SuperScript IV SuperScript IV MultiScribe MuLV
Optimal reaction temp 50°C 50°C 37°C
Reaction time 10 min 10 min 120 min
Ability to work with degraded or inhibitor-containing RNA High High Low
Reduced RNase H activity Yes Yes No
Primers included Random primers & oligo(dT)20 Random primers & oligo(dT)20 Random primers
Integrated gDNA removal No Yes No
Format All RT reaction components premixed All RT reaction components premixed RT reaction components in separate tubes
  Learn more Learn more Learn more
Other cDNA synthesis products for RT-qPCR
One-step RT-PCR kits for fast and simple workflow
  Superior choice for all RNAs, including difficult templates Value choice for standard applications
  SuperScript IV One-Step RT-PCR System SuperScript III One-Step RT-PCR System with Platinum Taq DNA Polymerase
Reverse transcriptase SuperScript IV SuperScript III
PCR enzyme Platinum SuperFi DNA DNA Polymerase Platinum Taq DNA Polymerase
Optimal reaction temperature 50-55°C 55°C
Reaction time 10 min 30 min
Sensitivity Superior Average
Maximum target length 13.8 kb 4.5 kb
  Learn more Learn more
Stand-alone enzymes for maximum flexibility in reaction setup
  Superior enzyme for all RNAs, including difficult templates Value enzyme for commonly used RNA templates
  Superscript IV Reverse Transcriptase M-MLV-Reverse Transcriptase
Optimal reaction temp 50–55°C 37°C
Reaction time 10 min 50 min
cDNA yield Superior Average
Sensitivity Superior Average
Max cDNA length Up to 12 kb Up to 7 kb
Ability to work with degraded or inhibitor-containing RNA High Low
Reduced RNase H activity Yes No
  Learn more Learn more
Other reverse transcriptases
Complete kits with all cDNA synthesis reaction components
  Superior choice for all RNAs, including difficult templates Value choice for commonly used RNA templates
  SuperScript IV
First-Strand Synthesis System
High-Capacity cDNA Reverse Transcription Kit
Reverse transcriptase SuperScript IV MultiScribe MuLV
Optimal reaction temp 50–55°C 37°C
Reaction time 10 min 50 min
cDNA yield Superior Average
Sensitivity Superior Average
Max cDNA length Up to 12 kb Up to 7 kb
Ability to work with degraded or inhibitor-containing RNA High Low
Reduced RNase H activity Yes No
  Learn more Learn more
Other cDNA synthesis kits for endpoint PCR applications
cDNA synthesis kits for the highest convenience in RT-qPCR setup
  Superior master mix for all RNA templates Superior master mix for all RNA templates, with fast and easy gDNA removal Value choice for commonly used RNA templates
  SuperScript IV VILO Master Mix SuperScript IV VILO Master Mix with ezDNase Enzyme High-Capacity cDNA Reverse Transcription Kit
Reverse transcriptase SuperScript IV SuperScript IV MultiScribe MuLV
Optimal reaction temp 50°C 50°C 37°C
Reaction time 10 min 10 min 120 min
Ability to work with degraded or inhibitor-containing RNA High High Low
Reduced RNase H activity Yes Yes No
Primers included Random primers & oligo(dT)20 Random primers & oligo(dT)20 Random primers
Integrated gDNA removal No Yes No
Format All RT reaction components premixed All RT reaction components premixed RT reaction components in separate tubes
  Learn more Learn more Learn more
Other cDNA synthesis products for RT-qPCR
One-step RT-PCR kits for fast and simple workflow
  Superior choice for all RNAs, including difficult templates Value choice for standard applications
  SuperScript IV One-Step RT-PCR System SuperScript III One-Step RT-PCR System with Platinum Taq DNA Polymerase
Reverse transcriptase SuperScript IV SuperScript III
PCR enzyme Platinum SuperFi DNA DNA Polymerase Platinum Taq DNA Polymerase
Optimal reaction temperature 50-55°C 55°C
Reaction time 10 min 30 min
Sensitivity Superior Average
Maximum target length 13.8 kb 4.5 kb
  Learn more Learn more
Need RNase Inhibitor to stabilize RNA molecules? Find out more
Featured videos
Resources
Education
Review educational resources for reverse transcription basics, cDNA synthesis, enzyme selection, troubleshooting tips, and cDNA applications.
Molecular Biology Webinar Series
View our webinar series to learn more about molecular biology topics such as reverse transcription, PCR, and cloning.
Mobile & Desktop Apps
Access scientific content and helpful tools through artis and Cloning Bench mobile apps.
OEM & customized cDNA solutions
Tailor-made performance tested products with a strict quality standards to meet your assay needs.
Support
PCR and cDNA Synthesis Support Center
Find tips, troubleshooting help, and resources for your endpoint PCR and cDNA applications.
Product Support Documentation
Search for manuals, protocols, Material Safety Data Sheets, product literature, and certificates by catalog number or product name.
Contact Us
Email or call our technical application scientists for additional questions regarding PCR enzymes and master mixes.