M-MLV Reverse Transcriptase (200 U/μL)
Invitrogen™

M-MLV Reverse Transcriptase (200 U/μL)

M-MLV Reverse Transcriptase is a recombinant DNA polymerase that synthesizes a complementary DNA strand from single-stranded RNA, DNA, or anRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
28025013200 Reactions
280250211,000 Reactions
Catalog number 28025013
Price (USD)
419.65
Online exclusive
456.00
Save 36.35
Each
No. of Reactions:
200 Reactions
Request bulk or custom format
Price (USD)
419.65
Online exclusive
456.00
Save 36.35
Each

M-MLV Reverse Transcriptase is a recombinant DNA polymerase that synthesizes a complementary DNA strand from single-stranded RNA, DNA, or an RNA:DNA hybrid. Compared to AMV RT, Moloney Murine Leukemia Virus Reverse Transcriptase (M-MLV RT) lacks DNA endonuclease activity and has a lower RNase H activity.

One unit of M-MLV RT is the amount of enzyme required to incorporate 1 nmole of deoxyribonucleotide into acid-precipitable material in 10 min at 37°C using poly(A) oligo(dT)25 as template-primer.

Features of M-MLV Reverse Transcriptase include:
• Excellent thermostability with optimal activity at 37°C
• Can be used to synthesize first-strand cDNA up to 7 kb
• Multiple applications including synthesis of first-strand cDNA, primer extension, sequencing dsDNA, cDNA libraries, and RT-PCR

Source
Purified from E. coli expressing the pol gene of M-MLV on a plasmid

Performance and quality testing
SDS-PAGE purity; endodeoxyribonuclease, exodeoxyribonuclease, and ribonuclease assays; and yield and length of cDNA product

Unit reaction conditions
50 mM Tris-HCl (pH 8.3), 40 mM KCl, 6 mM MgCl2, 1 mM DTT, 0.5 mM [3H]dTTP, 0.1 mM poly(A), 0.1 mM oligo(dT)25, 0.1 mg/mL BSA, and enzyme in 50 μL for 10 min at 37°C

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeFirst-Strand cDNA
FormatStand-alone Enzyme
No. of Reactions200 Reactions
Optimal Reaction Temperature37°C
Quantity40,000 units
Reaction FormatSeparate components
Reagent TypeReverse Transcription
Reverse TranscriptaseM-MLV
Ribonuclease H ActivityYes
Shipping ConditionWet Ice
Size (Final Product)Up to 7 kb
Starting MaterialRNA
TechniqueReverse Transcription
Concentration200 U/μL
Reaction Speed50 min.
Unit SizeEach
Contents & Storage

• M-mLV RT, 200 μL (200 U/μL)
• 5X First Strand Buffer, 1 mL
• DTT, 500 μL (100 mM)

Store at –20°C.

Frequently asked questions (FAQs)

How much of the first-strand cDNA reaction should I load for PCR?

While the volume is dependent on the starting amount of RNA used for the first-strand synthesis and the abundance of the target gene, we'd recommend starting with 10% of the first-strand reaction for your PCR reaction.

What is the highest temperature that MMLV, SuperScript II, SuperScript III, or SuperScript IV RTs can be used?

The optimal temperature for for MMLV is 42 degrees C. The optimal temperature for SuperScript II RT is 42 degrees C, and can be used up to 50 degrees C. The optimal temperature for SuperScript III RT is 50 degrees C, and can be used up to 55 degrees C. For some qRT-PCR reactions where gene-specific primers are used, you can do the RT reaction at 60 degrees C. The optimal temperature for SuperScript IV RT is 50 degrees C, but can be used up to 65 degrees C.

Can I use elevated temperatures for reverse transcription with Moloney Murine Leukemia Virus (M-MLV) reverse transcriptase?

Moloney Murine Leukemia Virus (M-MLV) Reverse Transcriptase is not well-suited for elevated temperatures. Typically, M-MLV RT operates optimally at temperatures around 37 degrees C to 42 degrees C.

To perform reverse transcription at elevated temperatures (e.g., 50 degrees C or higher), consider using thermostable reverse transcriptases such as SuperScript IV or Maxima H Minus, which are engineered to perform efficiently at higher temperatures.

Citations & References (4)

Citations & References
Abstract
Raloxifene Upregulated Mesangial Cell MMP-2 Activity via ER-ß Through Transcriptional Regulation.
Authors:Fang M, Wu XC, Huang W,
Journal:Cell Biochem Biophys
PubMed ID:23471663
'Raloxifene, a second-generation selective estrogen receptor modulator, exerts estrogen-like effects in specific tissues. In this present study, we examined the effect of raloxifene on mesangial cell matrix metalloproteinase-2 (MMP-2) activity in streptozotocin-induced diabetic mice. Raloxifene increased the MMP-2 level in a dose-dependent and receptor-mediated manner. An antibody against estrogen receptor-ß ... More
Single cell rt-PCR identification of odorant receptors expressed by olfactory neurons.
Authors:Malnic B,
Journal:Methods Mol Biol
PubMed ID:23585038
'Mammals have between 400 and 1,300 functional odorant receptor (OR) genes in their genomes. Each olfactory sensory neuron in the nose expresses only one single type of OR out of this vast repertoire. The OR expressed by an olfactory sensory neuron determines its functional activity and wiring to the olfactory ... More
Fast-mode duplex qPCR for BCR-ABL1 molecular monitoring: innovation, automation, and harmonization.
Authors:Gerrard G, Mudge K, Foskett P, Stevens D, Alikian M, White HE, Cross NC, Apperley J, Foroni L,
Journal:Am J Hematol
PubMed ID:22566190
Reverse transcription quantitative polymerase chain reaction (RTqPCR)is currently the most sensitive tool available for the routine monitoring of disease level in patients undergoing treatment for BCRABL1 associated malignancies. Considerable effort has been invested at both the local and international levels to standardise the methodology and reporting criteria used to assess ... More
Transforming growth factor-ß is required for vasculogenic mimicry formation in glioma cell line U251MG.
Authors:Ling G, Wang S, Song Z, Sun X, Liu Y, Jiang X, Cai Y, Du M, Ke Y,
Journal:Cancer Biol Ther
PubMed ID:22104964
Both vasculogenic mimicry (VM) and transforming growth factor-ß (TGFß) are positively correlated with malignancy in glioma. Accordingly, we supposed that TGFß might be related with VM, and aimed to detect whether TGFß could influence VM formation in two glioma cell lines U251MG and SHG44, which were different in malignancy. We ... More