Search
Whether you’re using a hard-to-transfect mammalian cell line, an animal model, or simply want efficient gene delivery, our ViraPower™ product portfolio is the ideal choice for either transient or stable expression. We offer two ViraPower™ systems—one for lentiviral expression and one for adenoviral expression. Both systems are are highly-efficient and offer reproducible delivery methods for RNAi or expression constructs. Both viral systems create replication-incompetent viral particles to help enable safe, effective delivery and expression of your gene or short hairpin RNA sequence in any mammalian cell type.
ViraPower™ Lentiviral Expression Systems stably integrate your gene of interest into your target cell's genome, making these systems ideal for functional studies. Our ViraPower™ HiPerform™ Expression Kits are all-inclusive kits that contain vectors with specialized elements that enable:
ViraPower™ Adenoviral Expression Systems deliver your gene of interest in trans for high level protein production. The ViraPower™ Adenoviral Gateway® Expression System quickly and easily provides high-level transient gene expression in any mammalian cell type.
| NEW | Protein Expression Support Center Find tips, troubleshooting help, and resources for your protein expression applications. |
For Research Use Only. Not for use in diagnostic procedures.