Request a Quote
32561 - MEM alpha, GlutaMAX(TM), no nucleosides
ComponentsMolecular WeightConcentration (mg/L)mM
Amino Acids
Glycine75.050.00.6666667
L-Alanine89.025.00.28089887
L-Alanyl-L-Glutamine217.0406.01.8709677
L-Arginine hydrochloride211.0105.00.49763033
L-Asparagine-H2O132.050.00.37878788
L-Aspartic acid133.030.00.22556391
L-Cysteine hydrochloride-H2O176.0100.00.5681818
L-Cystine240.024.00.1
L-Glutamic Acid147.075.00.5102041
L-Histidine155.031.00.2
L-Isoleucine131.052.40.4
L-Leucine131.052.40.4
L-Lysine146.058.00.39726028
L-Methionine149.015.00.10067114
L-Phenylalanine165.032.00.19393939
L-Proline115.040.00.3478261
L-Serine105.025.00.23809524
L-Threonine119.048.00.40336135
L-Tryptophan204.010.00.04901961
L-Tyrosine181.036.00.19889502
L-Valine117.046.00.3931624
Vitamins
Ascorbic Acid176.050.00.2840909
Biotin244.00.14.0983607E-4
Choline chloride140.01.00.007142857
D-Calcium pantothenate477.01.00.002096436
Folic Acid441.01.00.0022675737
Niacinamide122.01.00.008196721
Pyridoxal hydrochloride204.01.00.004901961
Riboflavin376.00.12.6595744E-4
Thiamine hydrochloride337.01.00.002967359
Vitamin B121355.01.360.0010036901
i-Inositol180.02.00.011111111
Inorganic Salts
Calcium Chloride (CaCl2-2H2O)147.0264.01.7959183
Magnesium Sulfate (MgSO4-7H2O)246.0200.00.8130081
Potassium Chloride (KCl)75.0400.05.3333335
Sodium Bicarbonate (NaHCO3)84.02200.026.190475
Sodium Chloride (NaCl)58.06800.0117.24138
Sodium Phosphate monobasic (NaH2PO4-2H2O)156.0158.01.0128205
Other Components
D-Glucose (Dextrose)180.01000.05.5555553
Lipoic Acid206.00.29.708738E-4
Phenol Red376.410.00.026567481
Sodium Pyruvate110.0110.01.0
Reference:
1. Simonsen, C.C. and Levinson, A.D. 1983. Isolation and expression of an altered mouse dihydrofolate reductase cDNA. Proc. Natl. Acad. Sci. U.S.A. 80:2495-2499.
Related Products:
41090 - MEM, GlutaMAX(TM)
Cat No: 41090028 , 41090036 , 41090093 , 41090101
42360 - MEM, HEPES, GlutaMAX(TM)
Cat No: 42360024 , 42360032 , 42360081 , 42360099
32571 - MEM alpha, GlutaMAX(TM), nucleosides
Cat No: 32571028 , 32571036 , 32571093 , 32571101