Mouse intestinal section visualized using fluorescent Qdot® nanocrystal conjugates.
A mouse intestinal section visualized using fluorescent Qdot® nanocrystal conjugates. Actin was labeled with a mouse anti-actin monoclonal antibody and visualized using red-fluorescent Qdot® 655 goat F(ab′)2 anti–mouse IgG (Q11022MP, Q11221MP). Laminin was labeled with a rabbit anti-laminin polyclonal antibody and visualized using green-fluorescent Qdot® 525 goat F(ab′)2 anti–rabbit IgG (Q11441MP). Nuclei were stained with blue-fluorescent Hoechst 33342 (H1399, H3570, H21492). Image contributed by Thomas Deerinck and Mark Ellisman, The National Center for Microscopy and Imaging Research, San Diego, CA.
A mouse intestinal section visualized using fluorescent Qdot® nanocrystal conjugates. Actin was labeled with a mouse anti-actin monoclonal antibody and visualized using red-fluorescent Qdot® 655 goat F(ab′)<sub>2</sub> anti–mouse IgG (Q11022MP, Q11221MP). Laminin was labeled with a rabbit anti-laminin polyclonal antibody and visualized using green-fluorescent Qdot® 525 goat F(ab′)<sub>2</sub> anti–rabbit IgG (Q11441MP). Nuclei were stained with blue-fluorescent Hoechst 33342 (H1399, H3570, H21492). Image contributed by Thomas Deerinck and Mark Ellisman, The National Center for Microscopy and Imaging Research, San Diego, CA.
Related Products
Related Images