Search
Find valuable information.
Optimize your experiments to get the best results. We’ve compiled a detailed knowledgebase of the top tips and tricks to meet your research needs.
View the relevant questions below:
Having problems with your experiment? Visit our
Yes, as long as the molecule of interest has the appropriate reactive group for the labeling products available. If you have a molecule that does not have a suitable reactive site compatible with the various reactive labeling products we offer, refer to texts titled “Handbook of Derivitization Methods for HPLC” for other labeling options.
After covalent modification, the function of the labeled molecule may be altered relative to the unlabeled molecule; the acceptable level of functionality and background should be determined empirically.
You must know something of the structure of your molecule, peptide, or protein to know what sites are suitable for labeling and that the intended site for labeling is accessible to the reactive labeling reagent. Avoid covalently modifying the active sites of enzymes or other strategically functional areas of the molecule. For example, if the carboxyl terminus of a peptide is required for binding or other function, avoid conjugating a dye to the carboxyl group or anywhere near the C-terminus.
No. For large molecules that may have more than one of the same reactive site for labeling, e.g., lysines on the surface of a protein, labeling with the amine-reactive reagent is ‘shotgun’ or random. It is not possible to control what sites will be conjugated, and the final product will be a mixed population of labeled protein with the label at different sites throughout the surface of the individual protein.
We highly recommend that you determine the DOL. DOL determination is achieved by absorbance readings at either 260 nm (for nucleic acids) or 280 nm (for proteins) plus the absorbance maximum for the fluorophore. It is only by an absorbance reading that you may determine if the labeling reaction was successful and, if subsequent labeling of the same molecule is to be done in the future, to establish the best level of labeling for that molecule.
Fluorescence detection cannot tell you if the molecule is unlabeled, under-labeled, or overlabeled— all of these labeling scenarios may provide the same level of little or no fluorescence. With some dyes, overlabeling may result in dye-dye quenching, resulting in no detectable fluorescence, but the dye will be detected using absorbance. Also, visual inspection of the color of the solution of the labeled molecule cannot determine if it was suitably labeled.
A DOL measurement may be performed for biotinylated-molecules using either the HABA assay or similar assays (see Cat. No. B30751 FluoReporter™ Biotin Quantitation Assay Kit).
Yes. Depending upon the reaction chemistry, labeling may be carried out in organic solvents such as DMSO, DMF, acetonitrile, alcohol, or other solvents. Useful information may be found in the text Bioconjugate Techniques (Greg T. Hermanson, Academic Press) or at Labeling Small Peptides with Amine-Reactive Dyes in Organic Solvents.
Reasons for the loss of signal from a fluorophore-conjugated molecule are varied, depending if the loss was due to the properties of the molecule or due to the fluorophore attached to the molecule. Poor signal may be due to poor binding or a higher on/off rate relative to the unlabeled molecule. Proteins or other molecules may be digested by proteases and the subsequent product comprising small, fluorophore-conjugated peptides or amino acids may become too diffuse for detection or wash away. Some fluorophore-conjugated molecules may be actively effluxed by live cells to a much greater extent than the unlabeled molecule.
Fluorophores attached to the probe of interest may become photobleached, quenched, or degraded. The presence of colorimetric dyes (e.g., phenol red, trypan blue), xylene, or other agents may transiently quench some fluorophores; once removed, the fluorophores are still fluorescent. Fluorophores may become irreversibly degraded by photobleaching, oxidation, reduction, or by other means. Avoid exposure to light during storage, the extremes of pH, strong oxidizing or reducing agents, heavy metals, and, for fluorescent proteins such as GFP or the phycobiliproteins (R-PE, APC), avoid anything that may promote digestion (i.e., by proteases), unfolding, or denaturation.
For Research Use Only. Not for use in diagnostic procedures.