    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ZNF81 Antibodies
          NeoBiotechnologies
          ZNF81/Zinc finger protein 81 (Transcription Factor) Monoclonal Antibody (PCRP-ZNF81-2C7)
          View all (8) ZNF81 antibodies
          Questions & Answers
          Cite ZNF81/Zinc finger protein 81 (Transcription Factor) Monoclonal Antibody (PCRP-ZNF81-2C7)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          ZNF81/Zinc finger protein 81 (Transcription Factor) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ZNF81/Zinc finger protein 81 (Transcription Factor) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          ZNF81/Zinc finger protein 81 (Transcription Factor) Antibody (347344-MSM1-P1) in ICC/IF
          Immunofluorescence analysis of PFA-fixed HeLa cells stained using ZNF81 Mouse Monoclonal Antibody (PCRP-ZNF81-2C7). followed by goat anti-mouse IgG-CF488 (green). CF640A phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ZNF81/Zinc finger protein 81 (Transcription Factor) Antibody in Immunocytochemistry (ICC/IF)
          ZNF81/Zinc finger protein 81 (Transcription Factor) Antibody in Flow Cytometry (Flow)
          ZNF81/Zinc finger protein 81 (Transcription Factor) Antibody in Peptide array (ARRAY)
          ZNF81/Zinc finger protein 81 (Transcription Factor) Antibody in SDS-PAGE (SDS-PAGE)
          ZNF81/Zinc finger protein 81 (Transcription Factor) Monoclonal Antibody (PCRP-ZNF81-2C7)
          Product Details
          347344-MSM1-P1
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1-2 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          1-2 µg/mL
          -
          Flow Cytometry (Flow)
          1-2 µg/10^6 cells
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          1-2 g/500 µg lysate
          -
          Peptide Array (Array)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          PCRP-ZNF81-2C7
          Immunogen
          Recombinant full-length human ZNF81 protein
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          200 µg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4, with 0.05% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          4°C, do not freeze
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          This gene encodes a protein that likely functions as a transcription factor. The protein contains an N-terminal KRAB domain and several C2H2-type zinc finger motifs. Mutations in this gene cause mental retardation X-linked type 45 (MRX45).
          Bioinformatics
          Protein Aliases: HFZ20; unnamed protein product; Zinc finger protein 81
          View more View less
          Gene Aliases: dJ54B20.6; HFZ20; MRX45; ZNF81
          View more View less
          UniProt ID: (Human) P51508
          View more View less
          Entrez Gene ID: (Human) 347344
          View more View less
          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding DNA binding zinc ion binding metal ion binding
          Process(es)
          regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline