    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • UPK3A Antibodies
          NeoBiotechnologies
          Uroplakin 3A (Bladder Carcinoma Marker) Polyclonal Antibody
          View all (15) UPK3A antibodies
          Questions & Answers
          Cite Uroplakin 3A (Bladder Carcinoma Marker) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Uroplakin 3A (Bladder Carcinoma Marker) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Uroplakin 3A (Bladder Carcinoma Marker) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Uroplakin 3A (Bladder Carcinoma Marker) Antibody (7380-RBP1-P1) in WB
          Western blot analysis in human HEK293 lysate in 1) absence and 2) presence of immunizing peptide using UPK3A Rabbit PAb. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Uroplakin 3A (Bladder Carcinoma Marker) Antibody in Western Blot (WB)
          Uroplakin 3A (Bladder Carcinoma Marker) Polyclonal Antibody
          Product Details
          7380-RBP1-P1
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1-2 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          A synthetic peptide, aa260-279 (GASESSYTSVNRGPPLDRAE) of human UPK3A
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          200 µg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.4, with 0.05% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          4°C, do not freeze
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          This gene encodes a member of the uroplakin family, a group of transmembrane proteins that form complexes on the apical surface of the bladder epithelium. Mutations in this gene may be associated with renal adysplasia. Alternatively spliced transcript variants have been described.
          Bioinformatics
          Protein Aliases: membrane protein; UP3a; UPIII; UPK3; uroplakin 3; Uroplakin III; Uroplakin-3a
          View more View less
          Gene Aliases: UP3A; UPIII; UPIIIA; UPK3; UPK3A
          View more View less
          UniProt ID: (Human) O75631
          View more View less
          Entrez Gene ID: (Human) 7380
          View more View less
          Function(s)
          protein binding
          Process(es)
          cell morphogenesis kidney development water transport urea transport epithelial cell differentiation potassium ion homeostasis sodium ion homeostasis urinary bladder development
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-jrc47:80/100.66.77.21:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline