    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
            • Aspire Member Program
            • Connect: Lab, Data, Apps
          • Primary Antibodies ›
          • SKIV2L2 Antibodies
          Bethyl Laboratories
          SKIV2L2 Polyclonal Antibody
          View all (8) SKIV2L2 antibodies
          Questions & Answers
          Cite SKIV2L2 Polyclonal Antibody
          • Antibody Testing Data (1)
          SKIV2L2 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          SKIV2L2 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          SKIV2L2 Antibody (A300-615A-T) in IP
          Detection of human SKIV2L2 by western blot of immunoprecipitates. Samples: Whole cell lysate (0.5 or 1.0 mg per IP reaction; 20% of IP loaded) from HeLa cells prepared using NETN lysis buffer. Antibodies: Affinity purified rabbit anti-SKIV2L2 antibody (Product # A300-615A) (lot 2) used for IP at 6 µg per reaction. For blotting immunoprecipitated SKIV2L2, rabbit anti-SKIV2L2 antibody A300-614A was used at 1 µg/mL. Detection: Chemiluminescence with an exposure time of 30 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SKIV2L2 Antibody in Immunoprecipitation (IP)
          SKIV2L2 Polyclonal Antibody
          Product Details
          A300-615A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residues 650 and 700 of Human SuperKiller Viralicidic Activity 2-Like 2.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Purification
          Antigen affinity chromatography
          Storage buffer
          tris citrate/phosphate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Based on 100% sequence identity, this antibody is predicted to react with Mouse
          Target Information
          SKIV2L2 may be involved in pre-mRNA splicing.
          Bioinformatics
          Protein Aliases: ATP-dependent helicase SKIV2L2; ATP-dependent RNA helicase DOB1; ATP-dependent RNA helicase SKIV2L2; Exosome RNA helicase MTR4; functional spliceosome-associated protein 118; Superkiller viralicidic activity 2-like 2; TRAMP-like complex helicase
          View more View less
          Gene Aliases: DOB1; fSAP118; KIAA0052; MTR4; MTREX; SKIV2L2
          View more View less
          UniProt ID: (Human) P42285
          View more View less
          Entrez Gene ID: (Human) 23517
          View more View less
          Function(s)
          RNA helicase activity protein binding ATP binding poly(A) RNA binding DNA metabolism protein
          Process(es)
          mRNA splicing, via spliceosome maturation of 5.8S rRNA rRNA processing RNA catabolic process
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States
          Your items have has been added!
          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline