    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
            • Aspire Member Program
            • Connect: Lab, Data, Apps
          • Primary Antibodies ›
          • BICD1 Antibodies
          Bethyl Laboratories
          BICD1 Polyclonal Antibody
          View all (5) BICD1 antibodies
          Questions & Answers
          Cite BICD1 Polyclonal Antibody
          • Antibody Testing Data (1)
          BICD1 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          BICD1 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          BICD1 Antibody (A303-334A-T) in IP
          Detection of human BICD1 by western blot of immunoprecipitates. Samples: Whole cell lysate (1 mg for IP, 20% of IP loaded) from Jurkat cells. Antibodies: Affinity purified rabbit anti-BICD1 antibody (Product # A303-334A) used for IP at 6 µg/mg lysate. BICD1 was also immunoprecipitated by rabbit anti-BICD1 antibody A303-335A, which recognizes a downstream epitope. For blotting immunoprecipitated BICD1, A303-335A was used at 1 µg/mL. Detection: Chemiluminescence with an exposure time of 30 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          BICD1 Antibody in Immunoprecipitation (IP)
          BICD1 Polyclonal Antibody
          Product Details
          A303-334A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 300 and 350 of human Bicaudal D Homolog 1.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Purification
          Antigen affinity chromatography
          Storage buffer
          tris citrate/phosphate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          BICD1 was initially identified as the human homolog to Drosophila Bicaudal-D protein that forms part of the cytoskeleton and mediates the correct sorting of mRNAs for oocyte- and axis-determining factors during oogenesis. BICD1 can bind to dynein-dynactin and co-localizes with Rab6A on the trans-Golgi network and cytoplasmic vesicles, suggesting BICD1 regulates COPI-independent Golgi-ER transport. BICD1 will also interact with Rab6B, a Rab6 isoform that is expressed primarily in brain. Live cell imaging showed bi-directional movement of Rab6B structures in SK-N-SH neurites, indicating that BICD1 is involved in the Rab6B regulation of retrograde cargo transport in neuronal cells.
          Bioinformatics
          Protein Aliases: Bic-D 1; bicaudal D homolog 1; cytoskeleton-like bicaudal D protein homolog 1; Protein bicaudal D homolog 1
          View more View less
          Gene Aliases: BICD; BICD1
          View more View less
          UniProt ID: (Human) Q96G01
          View more View less
          Entrez Gene ID: (Human) 636
          View more View less
          Function(s)
          structural constituent of cytoskeleton protein binding cytoskeletal adaptor activity Rab GTPase binding proteinase activated receptor binding dynactin binding dynein binding
          Process(es)
          RNA processing intracellular mRNA localization anatomical structure morphogenesis viral process protein localization to organelle stress granule assembly positive regulation of receptor-mediated endocytosis minus-end-directed organelle transport along microtubule microtubule anchoring at microtubule organizing center negative regulation of phospholipase C activity regulation of proteinase activated receptor activity negative regulation of phospholipase C-activating G-protein coupled receptor signaling pathway
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States
          Your items have has been added!
          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline