    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
            • Aspire Member Program
            • Connect: Lab, Data, Apps
          • Primary Antibodies ›
          • DDX42 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Bethyl Laboratories
          DDX42 Polyclonal Antibody
          View all (8) DDX42 antibodies
          Protocols
          Cite DDX42 Polyclonal Antibody
          DDX42 Polyclonal Antibody
          Product Details
          A303-354A-M
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:10,000
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 888 and 938 of human DEAD (Asp-Glu-Ala-Asp) Box Polypeptide 42.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Storage conditions
          4°C
          Shipping conditions
          Wet ice
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Based on 100% sequence identity, this antibody is predicted to react with Chicken and Orangutan
          Target Information
          This gene encodes a member of the Asp-Glu-Ala-Asp (DEAD) box protein family. Members of this protein family are putative RNA helicases, and are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. Two transcript variants encoding the same protein have been identified for this gene.
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States
          Your items have has been added!
          Host server : magellan-search-green-55658555d6-5jc54:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline