    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
            • Aspire Member Program
            • Connect: Lab, Data, Apps
          • Primary Antibodies ›
          • SAMD1 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Bethyl Laboratories
          Atherin Polyclonal Antibody
          View all (2) SAMD1 antibodies
          Protocols
          Cite Atherin Polyclonal Antibody
          Atherin Polyclonal Antibody
          Product Details
          A303-578A-M
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:10,000
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 488 and 538 of human Sterile Alpha Motif Domain Containing 1.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Storage conditions
          4°C
          Shipping conditions
          Wet ice
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          May play a role in atherogenesis by immobilizing LDL in the atherial wall.
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States
          Your items have has been added!
          Host server : magellan-search-green-55658555d6-72rkq:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline