    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
            • Aspire Member Program
            • Connect: Lab, Data, Apps
          • Primary Antibodies ›
          • SLAIN1 Antibodies
          Bethyl Laboratories
          SLAIN1 Polyclonal Antibody
          View all (10) SLAIN1 antibodies
          Questions & Answers
          Cite SLAIN1 Polyclonal Antibody
          • Antibody Testing Data (1)
          SLAIN1 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          SLAIN1 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          SLAIN1 Antibody (A303-644A-T) in IP
          Detection of human SLAIN1 by western blot of immunoprecipitates. Samples: Whole cell lysate (1 mg for IP, 20% of IP loaded) from HEK293T cells. Antibodies: Affinity purified rabbit anti-SLAIN1 antibody (Product # A303-644A) used for IP at 6 µg/mg lysate. SLAIN1 was also immunoprecipitated by rabbit anti-SLAIN1 antibody A303-645A, which recognizes a downstream epitope. For blotting immunoprecipitated SLAIN1, A303-645A was used at 1 µg/mL. Detection: Chemiluminescence with an exposure time of 30 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SLAIN1 Antibody in Immunoprecipitation (IP)
          SLAIN1 Polyclonal Antibody
          Product Details
          A303-644A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 450 and 500 of Human SLAIN Motif Family, Member 1.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Purification
          Antigen affinity chromatography
          Storage buffer
          phosphate/tris citrate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          SLAIN1 (SLAIN Motif Family Member 1) is a Protein Coding gene. An important paralog of this gene is SLAIN2. Microtubule plus-end tracking protein that might be involved in the regulation of cytoplasmic microtubule dynamics, microtubule organization and microtubule elongation.
          Bioinformatics
          Protein Aliases: SLAIN motif family, member 1; SLAIN motif-containing protein 1
          View more View less
          Gene Aliases: C13orf32; SLAIN1
          View more View less
          UniProt ID: (Human) Q8ND83
          View more View less
          Entrez Gene ID: (Human) 122060
          View more View less
          Function(s)
          protein binding
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States
          Your items have has been added!
          Host server : magellan-search-green-55658555d6-5jc54:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline