    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
            • Aspire Member Program
            • Connect: Lab, Data, Apps
          • Primary Antibodies ›
          • REXO2 Antibodies
          Bethyl Laboratories
          REXO2 Polyclonal Antibody
          View all (7) REXO2 antibodies
          Questions & Answers
          Cite REXO2 Polyclonal Antibody
          • Antibody Testing Data (2)
          REXO2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          REXO2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          REXO2 Antibody (A304-812A-T) in WB
          Detection of human REXO2 by western blot. Samples: Whole cell lysate (50 µg) from HeLa, HEK293T, and Jurkat cells prepared using NETN lysis buffer. Antibody: Affinity purified rabbit anti-REXO2 antibody (Product # A304-812A) (lot A304-812A-1) used for WB at 0.4 µg/mL. Detection: Chemiluminescence with an exposure time of 10 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          REXO2 Antibody in Western Blot (WB)
          REXO2 Antibody in Immunoprecipitation (IP)
          REXO2 Polyclonal Antibody
          Product Details
          A304-812A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:5,000
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 187 to 237 of Human Oligoribonuclease, mitochondrial.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Purification
          Antigen affinity chromatography
          Storage buffer
          phosphate/tris citrate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Based on 100% sequence identity, this antibody is predicted to react with Bovine
          Target Information
          Nucleases are components of DNA and RNA metabolism that carry out functions in DNA repair, replication, and recombination and in RNA processing and degradation. SFN is a homolog of Orn, a 3-prime-to-5-prime exoribonuclease of E. coli that attacks the free 3-prime hydroxyl group on single-stranded RNA, releasing 5-prime mononucleotides in a sequential manner.
          Bioinformatics
          Protein Aliases: C11orf61; Oligoribonuclease, mitochondrial; REX2, RNA exonuclease 2 homolog; RNA exonuclease 2 homolog; Small fragment nuclease
          View more View less
          Gene Aliases: CGI-114; REX2; REXO2; RFN; SFN; SMFN
          View more View less
          UniProt ID: (Human) Q9Y3B8
          View more View less
          Entrez Gene ID: (Human) 25996
          View more View less
          Function(s)
          3'-5'-exoribonuclease activity nucleic acid binding 3'-5' exonuclease activity
          Process(es)
          nucleobase-containing compound metabolic process nucleotide metabolic process RNA phosphodiester bond hydrolysis, exonucleolytic
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States
          Your items have has been added!
          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline