    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • IL-2 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          IL-2 Monoclonal Antibody (MQ1-17H12), Alexa Fluor™ 488
          View all (160) IL-2 antibodies
          Protocols
          Cite IL-2 Monoclonal Antibody (MQ1-17H12), Alexa Fluor™ 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          IL-2 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          IL-2 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          IL-2 Antibody (RHCIL220) in Flow
          Alexa Fluor® 488 of anti-human IL-2 mAb antibody stained intracellularly on IL-2 expressing commercially available human intracellular cytokine positive control cells (HiCK-1 cells), purchased from BD Biosciences. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          IL-2 Antibody in Flow Cytometry (Flow)
          Product Details
          RHCIL220
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          5 µL / 1x10^6 cells
          -
          Product Specifications
          Host/Isotype
          Rat / IgG2a, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          MQ1-17H12
          Immunogen
          Human IL-2.
          Conjugate
          Alexa Fluor™ 488 Alexa Fluor™ 488 Alexa Fluor™ 488
          Excitation/Emission Max
          499/520 nm View spectra spectra
          Form
          Liquid
          Storage conditions
          4°C
          Shipping conditions
          Wet ice
          RRID
          AB_10376174
          Target Information
          Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
          How to use the Panel Builder
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline