Search
Automate your entire sample preparation workflow with the EXTREVA ASE Accelerated Solvent Extractor. The EXTREVA ASE system combines and automates sample extraction, in-cell cleanup, and evaporation. Faster extraction, with four samples extracted in parallel, increases laboratory throughput and efficiency, and the patented gas-assisted extraction mechanism uses less solvent and produces less waste. Designed with components that minimize PFAS background, this system provides greater accuracy and sensitivity for PFAS and other POPs testing, ensuring you achieve more precise results in your analyses.
With the EXTREVA ASE system, prepare your solid and semi-solid samples for GC, GC-MS and LC-MS analyses with ease. Simply place your sample cells into the extraction rack, set the desired parameters on the user interface and let the system take care of the rest. When extraction is complete, the EXTREVA ASE system automatically transfers your extracts to the evaporation carousel, eliminating the need for manual transfer of samples and additional sample preparation instruments. No other instrument or manual method encompasses the entire workflow without requiring user interaction or sample transfer at each step. The EXTREVA ASE system provides true walk-away operation for the entire process, making it the perfect partner for your GC, GC-MS or LC-MS analysis.
Learn why the EXTREVA ASE system is the ideal sample preparation system to prepare samples for PFAS analysis to meet regulations like US EPA Method 1633A.
For labs that are extracting solid or semi-solid samples, the EXTREVA ASE system is the first fully automated sample-to-vial preparation platform. Common applications of the EXTREVA ASE system are to prepare samples for the analysis of:
The EXTREVA ASE system is thoughtfully designed to minimize PFAS background. This ensures more accurate and reliable results in PFAS analysis for environmental testing and compliance with local regulations. From environmental to food and beverage to pharma and biotech, the EXTREVA ASE system can be used in a variety of industries to prepare high-quality samples for GC- and LC-MS analysis.
Use ASE to accurately and efficiently meet the performance requirements of US EPA 1633A for PFAS analysis in soil samples.
Use the EXTREVA ASE system for fast, accurate extraction and evaporation of polycyclic aromatic hydrocarbons in soil samples.
Automate the entire sample preparation workflow for GC-ECD analysis of organochlorine pesticides (OCPs) in soil samples with the EXTREVA ASE system.
Efficiently extract plastic additives from pharmaceutical packaging using our new, fully automated parallel extraction and evaporation system.
The EXTREVA ASE system provides:
* Required field