Search
60–80M reads
~2.5 hours run time
| Throughput | Up to 15 Gb |
| Read length | Up to 200-base fragment reads |
| Number of reads passing filter | 60-80 million reads passing filter |
| Sequencing run time | 2.5 hours |
| Barcoding solutions | 384 barcodes supported by Torrent Suite Software 96 off-the-shelf barcodes for DNA 16 off-the-shelf barcodes for RNA |
| Software solutions | Torrent Suite Software utilizing Torrent Browser including TMAP alignment and Torrent Variant Caller for germline or somatic mutation detection. Torrent Browser (included) offers users remote web access to instrument status monitoring, run quality reports, individual data files, and extensible plug-ins for application-specific analysis. Plugins available for download from the Torrent Browser Plugin Store for analysis of genome sequencing, targeted sequencing, variant analysis and annotation, microbial sequencing, transfer to 3rd party bioinformatics packages, and more. Data can optionally flow into Ion Reporter Software for controlled analysis, annotation, and reporting of variants. Ion Reporter Software provides application specific workflows to support single, trio and paired tumor normal analysis. |
| Data formats | Industry standard FASTQ, SFF, BAM and VCF format outputs |
| Working environment (for indoor use only) | Temperature: 68-77° F (20-25° C) Humidity: 40-60%, noncondensing Altitude: <6,500 ft (2,000 m) Clearances:
12 in (30.5 cm) in rear
Optional rack mounting with two Ion Proton Sequencers per rack†4 in (10 cm) on left side 4 in (10cm) on right side 4 in (10 cm) from front edge of bench to sequencer bezel 36 in (90 cm) aisle in front of bench for operator access |
| Gas Supply | Connection: 0.25 in push-to-connect fitting Pressure: 30 psi Composition: nitrogen (grade 4.8, 99.998% or better) |
| Other connections | Ethernet: 1 GigE USB: 2x USB 2.0 |
| Power | Voltage: 100 V (min) to 240 V (max) Current: 14 A (max) Frequencing: 50/60 Hz Power Draw: 1,350 W |
| Dimensions | Width: 21.3 in/54.2 cm Depth: 30.5 in/77.5 cm Height: 18.7in/47.4 cm |
| Weight | Crated for shipment: 200 lb/90.7 kg Free-standing: 130 lb/59 kg |
†Space-saving rack can be purchased for the Ion Proton System. Contact your sales representative for details.
The trademarks mentioned herein are the property of Thermo Fisher Scientific Inc. and/or its affiliate(s) or their respective owners. Intel and Xeon are registered trademarks of Intel Corporation. NVIDIA and Tesla are registered trademarks of NVIDIA Corporation. Altera and Stratix are registered trademarks of of Altera Corporation. Ubuntu is a registered trademark of Canonical Ltd.
Our service plans help keep you up and running. With service and support that spans the sequencing workflow from sample prep through validation, our service plans give you peace of mind wherever your research takes you.
| Features | AB Complete | AB Assurance | AB Maintenance |
|---|---|---|---|
| Repair response time (days) | Guaranteed next day on-site repairs‡ | Guaranteed 2-day on-site repairs‡ | Target 2-day remedial repairs‡ |
| Remote instrument monitoring and diagnostics | ✓ | ✓ | |
| Priority phone and email access to instrument support | ✓ | ✓ | ✓ |
| Instrument operation software upgrades | ✓ | ||
| Instrument qualification services | ✓ | ||
| Planned maintenance | ✓ | ✓ | ✓ |
| Parts, labor and travel | Included | Included | Discount with Plus plan |
‡Availability limited in some geographic areas
We also offer bioinformatics services to help you harness the power of Ion Torrent technology.