For Clinicians
Resources for Medical Professionals  
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
  Food  
  Seasonal  
  Indoor  
  Allergic Asthma  
Is your patient a candidate for specific IgE testing?
Find out 
Get detailed information on whole allergens and allergen components. 
Allergen Encyclopedia 
Ready to test a patient?
On-Demand Education 
Boxelder tree
Allergy Facts, Symptoms, and Treatment
Boxelder trees, which are also known as ash leaf maples and Manitoba maples, are native to North America and parts of Central America; however, the species also has been naturalized in Europe. Due to their quick growth and ability to provide shade, they're often planted in urban environments.1 While all maple species are potentially allergenic, boxelder has significantly allergenic pollen and is wind pollinated.2
Where are boxelder trees found?
While the boxelder has become naturalized in Europe, its native range covers the East Coast of the United States to California and from Alberta to southern Mexico and Guatemala. 1
Prepare for your next visit with your healthcare provider. 
Fill out our symptom tracker
Learn about specific IgE allergy testing options.
Read more
Are there other allergens I could be sensitized to?*
Many patients with boxelder tree allergy can experience symptoms when exposed to other allergens such as tree, weed, or grass pollens, making it difficult to determine which pollen is causing the symptoms, especially when pollen seasons are overlapping. This is called cross-reactivity and occurs when your body's immune system identifies the proteins, or components, in different substances as being structurally similar or biologically related, thus triggering a response. Other respiratory allergens that may cause reactions associated with boxelder pollen allergy are maples and other tree, weed, and grass pollens.7,8
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
How do I manage my allergy?
The management of allergic rhinitis includes avoidance of relevant allergens, symptomatic treatment, and allergen immunotherapy.4-6
  • Check local pollen counts daily and limit time outside when tree-pollen counts are high. Rain helps clear pollen from the air, so the best time to go outside is after a good rain. 
  • Delegate outdoor chores whenever possible and wear a pollen mask if you must do outside tasks. 
  • Keep windows closed and use air conditioning instead.
  • Wash bedding at least once a week in hot, soapy water.
  • Wash your clothes after outdoor activities and dry all clothes in a dryer as opposed to line drying outdoors.
  • Bathe and wash your hair every day before bedtime to keep pollen out of your bed.
  • Wipe off any pets to remove pollen before letting them into your home.
  • Ensure everyone removes their shoes before entering your home.
  • Use certified asthma and allergy air filters.
  • Pharmacological treatment, including antihistamines, corticosteroids, decongestants, and saline douches.
  • Allergen immunotherapy as directed by your healthcare provider.
Looking for more allergy info and management tips?
Visit the Living with Allergies section
Common Symptoms
Boxelder tree allergy symptoms can be similar to many other pollen allergies and may include:3,4
  • Sneezing
  • Nasal congestion
  • Runny nose
  • Watery eyes
  • Itchy throat and eyes
  • Wheezing
If you're sensitized to boxelder trees and have asthma, tree pollen may trigger or worsen asthma symptoms, such as coughing and wheezing.3,4
How do I know if I'm allergic?*
Together with your symptom history, skin-prick testing or specific IgE blood testing can help determine if you are sensitized to a particular allergen. If you are diagnosed with an allergy, your healthcare provider will work with you to create a management plan.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
Pollen Season
While tree pollen is common in the spring, boxelders typically produce pollen between March and May. 1,3
  1. USDA Plants Database [Internet].|place unknown|:United States Department of Agriculture; 2001 Jan 17 [2003 Feb 7] [2006 May 6]. Available from: https://plants.usda.gov/plantguide/pdf/pg_acne2.pdf.
  2. Pollen.com [Internet]. Plymouth Meeting, PA: IQVIA Inc.; 2020. Available from: https://www.pollen.com/research/genus/acer.
  3. American College of Allergy, Asthma & Immunology [Internet]. Arlington Heights, IL: American College of Allergy, Asthma & Immunology; 2014 [2018 Apr 23]. Available from: https://acaai.org/allergies/types/pollen-allergy.
  4. Roberts, Graham & Xatzipsalti, M & Borrego, Luis & Custovic, Adnan & Halken, Susanne & Hellings, Peter & Papadopoulos, Nikolaos & Rotiroti, G & Scadding, Glenis & Timmermans, Frans & Valovirta, Erkka. (2013). Paediatric rhinitis: Position paper of the European Academy of Allergy and Clinical Immunology. Allergy. 68. 10.1111/all.12235.
  5. Asthma and Allergy Foundation of America [Internet]. Arlington, VA: Asthma and Allergy Foundation of America; 2019 Apr 9. Available from: https://community.aafa.org/blog/tips-for-preventing-allergic-reactions-to-tree-and-grass-pollen.
  6. Mayo Clinic [Internet]. Rochester, MN; Mayo Foundation for Medical Education and Research; 2020 Apr 16. Available from: https://www.mayoclinic.org/diseases-conditions/hay-fever/in-depth/seasonal-allergies/art-20048343.
  7. Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Hilger C, Hofmaier S, Aalberse RC, Agache I, Asero R, Ballmer-Weber B, Barber D, Beyer K, Biedermann T, Bilò MB, Blank S, Bohle B, Bosshard PP, Breiteneder H, Brough HA, Caraballo L, Caubet JC, Crameri R, Davies JM, Douladiris N, Ebisawa M, EIgenmann PA, Fernandez-Rivas M, Ferreira F, Gadermaier G, Glatz M, Hamilton RG, Hawranek T, Hellings P, Hoffmann-Sommergruber K, Jakob T, Jappe U, Jutel M, Kamath SD, Knol EF, Korosec P, Kuehn A, Lack G, Lopata AL, Mäkelä M, Morisset M, Niederberger V, Nowak-W?grzyn AH, Papadopoulos NG, Pastorello EA, Pauli G, Platts-Mills T, Posa D, Poulsen LK, Raulf M, Sastre J, Scala E, Schmid JM, Schmid-Grendelmeier P, van Hage M, van Ree R, Vieths S, Weber R, Wickman M, Muraro A, Ollert M. EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol. 2016 May;27 Suppl 23:1-250. doi: 10.1111/pai.12563. PMID: 27288833.
  8. Dales RE et.al. Tree pollen and hospitalization for asthma in urban Canada. Int Arch Allergy Immunol. 2008;146(3):241-7.