Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
October 2022 | Linda Armstrong | ✓ Medically reviewed by: Gary Falcetano, PA-C, AE-C; Fabio Iachetti, MD; Eva Södergren, PhD, MSc
A licensed Physician Assistant with more than 25 years of diverse experience in emergency and disaster medicine, primary care, and allergy and immunology, Gary Falcetano is the U.S. Clinical Affairs Manager for Allergy in ImmunoDiagnostics at Thermo Fisher Scientific. Fabio Iachetti is a licensed physician with more than 15 years of diverse experience in several disease areas such as allergy, CV, pain, GI, rheumatology, urology, and diabetology. He is a Senior Medical Manager for Allergy in ImmunoDiagnostics Global Medical Affairs at Thermo Fisher Scientific. A nutritionist by training, Eva Södergren now works as a Senior Scientific Advisor for Allergy on the Medical and Scientific Affairs team for Thermo Fisher Scientific’s ImmunoDiagnostics division.
There are several different options for allergy testing, including blood tests, skin-prick tests (SPTs), and food challenge tests. Together with your healthcare provider, you can decide which test is best for you. After your elected allergy test is performed, the results are reviewed by your healthcare provider alongside your medical history to help establish an accurate diagnosis.
A blood test to aid allergy diagnosis is a quick and simple way for your healthcare provider to get answers to your underlying allergy questions. The test, also called a specific IgE (sIgE) blood test, provides informative results that indicate the levels of allergen-specific IgE antibodies in the blood. These antibodies are an indicator of allergic sensitization and, in context of an allergy-focused medical history, can help your healthcare provider determine if you are allergic and to what.
This powerful diagnostic tool can reveal potential sensitization to hundreds of possible allergens. Blood testing has fewer limitations on who can receive it or when it can be performed compared to other types of testing. Learn more about the benefits of specific IgE blood testing. Or explore a list of frequently asked questions.
There are two broad types of specific IgE blood tests: whole allergen tests and allergen component tests. Paired with a symptom history and physical exam, whole allergen tests help your healthcare provider to determine if you’re sensitized to a specific allergen or group of allergens.
Meanwhile, testing with allergen components helps providers identify the specific molecules (e.g., proteins, carbohydrates, enzymes) that may cause reactions. Knowing which molecules you’re sensitized to may help your provider determine if there could be an increased risk for a severe systemic reaction.
Additionally, testing with allergen components might reveal to providers that you can tolerate an allergen in a specific form. For example, if you have an egg allergy, this testing may suggest that you’re likely to tolerate baked eggs but not lightly cooked or raw eggs. (That means you may be able to eat baked goods such as sweets, breads, and more.)
Those healthcare providers who have used blood tests for decades may be most familiar with the term radioallergosorbent test (RAST), which measures specific IgE but is actually a term for an older technology. In fact, it doesn’t represent modern specific IgE blood tests for allergies, and any literature about RAST likely doesn’t apply to current tests.
With SPTs, your skin is directly exposed to suspected allergens and observed for signs of a reaction.
Specific IgE tests and SPTs are frequently used to help diagnose allergies. But how do you and your healthcare provider choose between them? Here are some key differences followed by a handy chart to further highlight the comparison.
Key Test Differences
Specific IgE Blood Test
SPT
No Risk of Anaphylaxis
✓
May be Performed by Most Healthcare Providers
✓
Same Day Results
✓
Not Affected by Medications
✓
Not Impacted by Skin Pigmentation/Condition
✓
Effective for People of Any Age
✓
A challenge test, in connection with a blood test or an SPT, is a powerful diagnostic tool. Also called an oral food challenge (OFC), it is used to establish a correct diagnosis, which can be guided by the results from an SPT or a blood test. A challenge test can be used to confirm a food allergy or to determine whether you have outgrown a food allergy. 8
An OFC typically has three steps:8
To properly manage and treat allergies, it’s important to first understand exactly what is causing symptoms. Talk to a healthcare provider about allergy testing options. To get the conversation started, we compiled a few questions designed to help healthcare providers get a full picture of your symptoms. You can review your answers together during your office visit.
Track allergy symptoms and prepare for a visit with a healthcare provider.
Learn about specific allergens, including common symptoms, management, and relief.
Are you a healthcare provider? Get comprehensive information on hundreds of whole allergens and allergen components.