FIGURE: 1 / 4
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
This affinity purified antibody is directed against human Ephrin B1, B2, B3. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with Ephrin-B1 to human, mouse, rat, and Gallus gallus; 100% homology with Ephrin-B2 to mouse; 100% homology with Ephrin-B3 to human and mouse.
Eph receptors and ephrins exhibit complementary expression in many tissues during embryogenesis indicating that bidirectional activation of Eph receptors and ephrin-B proteins may occur at expression domain interfaces. Ephrin-B1 transduces outside-in signals through C-terminal protein interactions that effect integrin-mediated cell attachment and migration. The distribution of ephrin-B1 in the developing retina suggests that it influences retinal axon mapping along the dorsal-ventral axis and may be involved in intratectal development The transmembrane ligand ephrin-B2 and its receptor tyrosine kinase EphB4 are specifically expressed on arterial and venous endothelial cells, respectively.
Protein Aliases: CEK5 receptor ligand; CEK5-L; EFL-3; ELF-2; ELK ligand; ELK-L; EPH-related receptor transmembrane ligand ELK-L3; EPH-related receptor tyrosine kinase ligand 2; EPH-related receptor tyrosine kinase ligand 5; EPH-related receptor tyrosine kinase ligand 8; Ephrin-B1; Ephrin-B2; Ephrin-B3; HTK ligand; HTK-L; LERK-2; LERK-5; LERK-8; ligand of eph-related kinase 5; ligand of the eph-related receptor tyrosine kinases hek and elk; Stimulated by retinoic acid gene 1 protein; unnamed protein product
Gene Aliases: Cek5-L; CFND; CFNS; EFB1; EFL-3; EFL-6; EFL3; EFL6; EFNB1; EFNB2; EFNB3; ELF-2; ELF-3; Elf2; Elk-L; Elk-L3; ephrin-B2; Epl2; Epl5; Epl8; EPLG2; EPLG5; EPLG8; Htk-L; HTKL; LERK-2; LERK-5; LERK-8; LERK2; LERK5; LERK8; NLERK-1; NLERK-2; Stra1
UniProt ID: (Human) P98172, (Human) P52799, (Human) Q15768, (Mouse) P52795, (Mouse) P52800, (Mouse) O35393
Entrez Gene ID: (Human) 1947, (Human) 1948, (Human) 1949, (Mouse) 13641, (Mouse) 13642, (Mouse) 13643