    • 엔터프라이즈급 실험실 정보 처리
    • 360° CDMO 및 CRO 서비스
    • CDMO 서비스
    • 임상시험 CRO 서비스
    • 장비 서비스
    • 교육 서비스
    • Unity Lab Services
    • 서비스 전체 보기
  • 지원
    • 고객센터
    • 문의하기
    • 시험성적서(COA) 및 적합성 인증서(COC)
    • Safety Data Sheets (SDS)
    • 매뉴얼
    • 인용 및 참고 문헌
    • 기기 지원
    • 기술 자료/제품 FAQ
    • 학습 센터
    • 지원 메뉴 전체보기
      Cell Culture Plastics
  • 문의하기
  • 빠른 주문
  • 주문 현황
  • 제품 문서 검색
Thermo Fisher Scientific Logo
Search
전체검색
Search button
          • 주문 현황
          • 빠른주문
          • 로그인
            로그인
            회원이 아니신가요? 계정 생성하기
            • 계정정보
            • 주문내역조회
            • 커스텀제품 및 프로젝트
            • Services Central
          • Primary Antibodies ›
          • GPNMB Antibodies
          Invitrogen
          GPNMB Recombinant Rat Monoclonal Antibody (CTSREVL), Alexa Fluor™ Plus 647
          Advanced Verification
          View all (45) GPNMB antibodies
          Tech Support
          Cite GPNMB Recombinant Rat Monoclonal Antibody (CTSREVL), Alexa Fluor™ Plus 647
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          • Advanced Verification (2)
          GPNMB Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          GPNMB Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          GPNMB Antibody (740113TP647) in ICC/IF
          Immunofluorescence analysis of GPNMB was performed using GPNMB Recombinant Rat Monoclonal Antibody (CTSREVL), Alexa Fluor™ Plus 647 (Product # 740113TP647) on 70-80% confluent B16-F10 and EL4 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes and then blocked with 2% BSA for 1 hour at room temperature. The cells were stained with GPNMB Recombinant Rat Monoclonal Antibody (CTSREVL), Alexa Fluor™ Plus 647 (Product # 740113TP647, 1:100) in 0.1% BSA, incubated at 4 degree Celsius overnight, Panel a) shows representative image ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GPNMB Antibody in Immunocytochemistry (ICC/IF)
          GPNMB Antibody in Flow Cytometry (Flow)
          GPNMB Antibody in Flow Cytometry (Flow)
          GPNMB Antibody
          GPNMB Antibody
          Product Details
          740113TP647
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:100
          -
          Flow Cytometry (Flow)
          0.125 µg/test
          -
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Rat / IgG2a, kappa
          Expression System
          Expi293
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          CTSREVL
          Conjugate
          Alexa Fluor™ Plus 647 Alexa Fluor™ Plus 647 Alexa Fluor™ Plus 647
          • Unconjugated
          • Alexa Fluor Plus 488 
          • Alexa Fluor Plus 555 
          • Request custom conjugation
          Excitation/Emission Max
          658/675 nm View spectra spectra
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          50 µg
          Purification
          Affinity chromatography
          Storage buffer
          proprietary buffer, pH 6.3-7.3
          Contains
          0.008% Bromonitrodioxane, 0.008% Methylisothiazolone
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Wet ice
          Product Specific Information
          Alexa Fluor™ Plus recombinant antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning data. Alexa Fluor™ Plus antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor™ Plus antibodies provide brighter signal compared to leading Alexa Fluor™ antibodies, providing you with better signal-to-noise for your critical experiments. These antibodies show better specificity and lot-to-lot consistency as these are recombinant antibodies, generated by cloning specific genes for the desired antibodies into an expression vector and expressed in vitro.
          Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining.
          It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
          Excitation: 658 nm; Emission: 675 nm; Laser: Red Laser
          Filtration: 0.2 µm post-manufacturing filtered.
          Target Information
          The protein encoded by this gene is a type I transmembrane glycoprotein which shows homology to the pMEL17 precursor, a melanocyte-specific protein. GPNMB shows expression in the lowly metastatic human melanoma cell lines and xenografts but does not show expression in the highly metastatic cell lines. GPNMB may be involved in growth delay and reduction of metastatic potential. Two transcript variants encoding different isoforms have been found for this gene.
          Bioinformatics
          Protein Aliases: DC-HIL; dendritic cell-associated heparin sulfate proteoglycan-dependent integrin ligand; Dendritic cell-associated transmembrane protein; iris pigment dispersion; Osteoactivin; Transmembrane glycoprotein NMB
          View more View less
          Gene Aliases: DC-HIL; Dchil; Gpnmb; Hgfin; ipd; Nmb
          View more View less
          UniProt ID: (Mouse) Q99P91
          View more View less
          Entrez Gene ID: (Mouse) 93695
          View more View less
          Function(s)
          integrin binding heparin binding membrane-bound signaling molecule
          Process(es)
          cell adhesion
          Contact tech support
          온라인 주문 Plus Icon Minus Icon
          • 주문 현황
          • 주문 지원
          • 빠른 주문
          • Supply Center
          • eProcurement
          지원 Plus Icon Minus Icon
          • Help and Support
          • 고객 센터
          • 기술 지원 센터
          • 제품 문서 검색
          • 사이트 문제 보고
          교육 및 이벤트 Plus Icon Minus Icon
          • 교육 센터
          • 프로모션
          • 이벤트 및 웨비나
          • 소셜 미디어
          • 소개 소개
          • 채용 채용
          • 투자자 투자자
          • 뉴스 뉴스
          • 사회적 책임 사회적 책임
          • Trademarks
          • 공정거래 공정거래
          • 정책 및 고지
          • PPD
          • 이용 약관
          • 개인정보 처리방침
          • 가격 및 운임 정책
          Korea flag icon
          Korea
          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat
          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat
          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat
          써모 피셔 사이언티픽 코리아 주식회사
          대표자 : 석수진
          사업자 등록번호 : 117-81-46910
          입금계좌 : 하나은행 336-890014-06204
          (예금주 : 써모피셔사이언티픽코리아 주식회사)
          써모 피셔 사이언티픽 솔루션스 유한회사
          대표자 : 석수진
          사업자 등록번호 : 114-86-04783
          입금계좌 : 신한은행 140-004-396660
          (예금주 : 써모피셔사이언티픽솔루션스 유한회사)
          주소 : 서울시 강남구 광평로 281 수서오피스빌딩 12층 06349 | 통신판매업신고번호 : 2015-서울강남-00898
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline