Sequence of this protein is as follows: D/EFTWHRRY GHGVSEEDKC
Contactin 1 encodes a protein that is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Protein Aliases: Contactin-1; Glycoprotein gp135; Neural cell surface protein F3; unnamed protein product
Gene Aliases: AW495098; CMYO12; CMYP12; CNTN; CNTN1; F3; F3cam; GP135; MYPCN
UniProt ID: (Human) Q12860, (Rat) Q63198, (Mouse) P12960
Entrez Gene ID: (Human) 1272, (Rat) 117258, (Mouse) 12805