    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • Contactin 1 Antibodies
          Antibodies Inc
          Contactin/F3 Monoclonal Antibody (K73/20), FL650
          View all (17) Contactin 1 antibodies
          Tech Support
          Cite Contactin/F3 Monoclonal Antibody (K73/20), FL650
          Additional Information:
          {{banner.description}}
          Contactin/F3 Monoclonal Antibody (K73/20), FL650
          Product Details
          75-038-FL650
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          K73/20
          Immunogen
          Synthetic peptide amino acids 21-37 of rat contactin (accession number A57112).
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: D/EFTWHRRY GHGVSEEDKC
          Target Information
          Contactin 1 encodes a protein that is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
          Bioinformatics
          Protein Aliases: Contactin-1; Glycoprotein gp135; Neural cell surface protein F3; unnamed protein product
          View more View less
          Gene Aliases: AW495098; CMYO12; CMYP12; CNTN; CNTN1; F3; F3cam; GP135; MYPCN
          View more View less
          UniProt ID: (Human) Q12860, (Rat) Q63198, (Mouse) P12960
          View more View less
          Entrez Gene ID: (Human) 1272, (Rat) 117258, (Mouse) 12805
          View more View less
          Function(s)
          protein binding carbohydrate binding protein binding involved in cell-cell adhesion glycoprotein binding
          Process(es)
          cell adhesion Notch signaling pathway positive regulation of gene expression positive regulation of sodium ion transport positive regulation of neuron projection development cerebellum development neuron projection development positive regulation of peptidyl-tyrosine phosphorylation axon guidance brain development locomotory behavior gene expression central nervous system myelin formation myelination cell-cell adhesion
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline