    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Contactin 1 Antibodies
          Antibodies Inc
          Contactin/F3 Monoclonal Antibody (K73/20), FL550
          View all (22) Contactin 1 antibodies
          Tech Support
          Cite Contactin/F3 Monoclonal Antibody (K73/20), FL550
          Additional Information:
          {{banner.description}}
          Contactin/F3 Monoclonal Antibody (K73/20), FL550
          Product Details
          75-038-FL550
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          K73/20
          Immunogen
          Synthetic peptide amino acids 21-37 of rat contactin (accession number A57112).
          View immunogen
          Conjugate
          FL550 FL550 FL550
          • Unconjugated
          • FL490
          • FL594
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          550/575 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: D/EFTWHRRY GHGVSEEDKC
          Target Information
          Contactin 1 encodes a protein that is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
          Bioinformatics
          Protein Aliases: Contactin-1; Glycoprotein gp135; Neural cell surface protein F3; unnamed protein product
          View more View less
          Gene Aliases: AW495098; CMYO12; CMYP12; CNTN; CNTN1; F3; F3cam; GP135; MYPCN
          View more View less
          UniProt ID: (Human) Q12860, (Rat) Q63198, (Mouse) P12960
          View more View less
          Entrez Gene ID: (Human) 1272, (Rat) 117258, (Mouse) 12805
          View more View less
          Function(s)
          protein binding carbohydrate binding protein binding involved in cell-cell adhesion glycoprotein binding
          Process(es)
          cell adhesion Notch signaling pathway positive regulation of gene expression positive regulation of sodium ion transport positive regulation of neuron projection development cerebellum development neuron projection development positive regulation of peptidyl-tyrosine phosphorylation axon guidance brain development locomotory behavior gene expression central nervous system myelin formation myelination cell-cell adhesion
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline