    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Ataxin-1 (11NQ) Antibodies
          Antibodies Inc
          Ataxin-1/11NQ Monoclonal Antibody (N76/8), FL594
          View all (10) Ataxin-1 (11NQ) antibodies
          Tech Support
          Cite Ataxin-1/11NQ Monoclonal Antibody (N76/8), FL594
          Additional Information:
          {{banner.description}}
          Ataxin-1/11NQ Monoclonal Antibody (N76/8), FL594
          Product Details
          75-117-FL594
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N76/8
          Immunogen
          Synthetic peptide amino acids 164-197 of mouse ataxin-1 (accession number P54254).
          3D Epitope / Immunogen
          Conjugate
          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          594/615 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: ATTPSQRSQL EAYSTLLANM GSLSQAPGHK VEPP
          Target Information
          Ataxin-1 (11NQ) refers to a specific isoform or variant of the Ataxin-1 protein, characterized by having 11 normal glutamine (Q) residues in its polyglutamine (polyQ) tract. This is in contrast to the expanded polyQ tracts found in disease-causing variants associated with spinocerebellar ataxia type 1 (SCA1). Ataxin-1 (11NQ) serves as a model for studying the normal physiological roles of Ataxin-1 and provides a contrast to the pathogenic forms associated with neurodegenerative diseases, enhancing our understanding of protein misfolding and aggregation disorders.
          Bioinformatics
          Protein Aliases: Alt-ATXN1; alternative ataxin1; Ataxin-1; spinocerebellar ataxia 1; spinocerebellar ataxia 1 homolog; Spinocerebellar ataxia type 1 protein; Spinocerebellar ataxia type 1 protein homolog
          View more View less
          Gene Aliases: 2900016G23Rik; ATX1; ATXN1; C85907; D6S504E; ENSMUSG00000074917; Gm10786; SCA1
          View more View less
          UniProt ID: (Human) P54253, (Mouse) P54254, (Rat) Q63540
          View more View less
          Entrez Gene ID: (Human) 6310, (Mouse) 20238, (Rat) 25049
          View more View less
          Function(s)
          DNA binding chromatin binding RNA binding protein binding poly(U) RNA binding POZ domain binding poly(G) binding identical protein binding protein C-terminus binding protein self-association molecular_function
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter transcription, DNA-templated regulation of transcription, DNA-templated adult locomotory behavior visual learning negative regulation of phosphorylation negative regulation of insulin-like growth factor receptor signaling pathway negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter lung alveolus development nuclear export excitatory postsynaptic potential positive regulation of glial cell proliferation biological_process transcription from RNA polymerase II promoter RNA processing brain development learning memory social behavior insulin-like growth factor receptor signaling pathway
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline