FIGURE: 1 / 1
For maximum recovery of product, centrifuge the vial prior to removing the cap.
CDY1, CDY2, CDYL1, and CDYL2 are members of the chromodomain Y-like (CDY) family of proteins, which are involved in chromatin remodeling and gene regulation. CDY1 and CDY2 are Y-linked genes specifically important for spermatogenesis, while CDYL1 and CDYL2 are autosomal and have broader roles in regulating chromatin structure and gene expression in various tissues. These proteins contribute to processes such as development, differentiation, and cellular response to environmental signals, and their dysfunction can be linked to reproductive and developmental disorders.
Protein Aliases: CDY1; CDY1/CDY2A/CDY1L1/CDYL2; CDY2; chromodomain protein, Y chromosome, 1; chromodomain protein, Y chromosome, 2; chromodomain protein, Y-linked, 1; chromodomain protein, Y-linked, 2A; testis-specific chromodomain protein on Y; Testis-specific chromodomain protein Y 1; Testis-specific chromodomain protein Y 2; unnamed protein product; Y chromosome chromodomain protein 2A
Gene Aliases: CDY; CDY1; CDY1A; CDY1B; CDY2; CDY2A; CDY2B
UniProt ID: (Human) Q9Y6F8, (Human) Q9Y6F7
Entrez Gene ID: (Human) 9085, (Human) 9426